Nitronium tetrafluoroborate, 0.5M solution in sulfolane
Nitronium tetrafluoroborate, 0.5M solution in sulfolane
Nitronium tetrafluoroborate, 0.5M solution in sulfolane
Nitronium tetrafluoroborate, 0.5M solution in sulfolane
Nitronium tetrafluoroborate, 0.5M solution in sulfolane
Thermo Scientific Chemicals

Nitronium tetrafluoroborate, 0.5M solution in sulfolane

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
200 mL
Packaging:
Glass bottle
Catalog number 175132000
Price (EUR)
627,00
Each
Quantity:
200 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
627,00
Each
Chemical Identifiers
CAS13826-86-3
IUPAC Namedioxoamine; tetrafluoroboranuide
Molecular FormulaBF4NO2
InChI KeyXYRQURYIJKJVSF-UHFFFAOYSA-N
SMILES[O-][NH+]=O.F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Solution
Strength0.48 to 0.55 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only