3-Bromofuran, 98%, stabilized with CaCO3
3-Bromofuran, 98%, stabilized with CaCO3
3-Bromofuran, 98%, stabilized with CaCO3
Thermo Scientific Chemicals

3-Bromofuran, 98%, stabilized with CaCO3

CAS: 22037-28-1 | C4H3BrO | 146.97 g/mol
Have Questions?
Catalog NumberQuantityPackaging
17526010010 mLGlass bottle
Catalog number 175260100
Price (USD)
-
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS22037-28-1
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4955 to 1.4985 (20°C, 589 nm)
Stabilizer0.5 % calcium carbonate
Appearance (Color)Clear colorless to brown
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only