DRI™ Gravity-Detect™ Specimen Validity Test Controls
Thermo Scientific™
DRI™ Gravity-Detect™ Specimen Validity Test Controls
Combat the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
17571.030 μg/mL
17561.015 μg/mL
Catalog number 1757
Price (USD)
-
Concentration:
1.030 μg/mL
Combat the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Gravity-Detect™ Specimen Validity Test Controls. Liquid and ready-to-use, these controls can be performed on a variety of general chemistry analyzers. The use of additives and adulterants to mask controlled substances in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution.
Our adulteration assays provide a complete and automated way to test for sample integrity and adulteration.
  • Associated calibrators and controls offered in convenient sizes
  • Liquid, ready-to-use
  • Can be performed on a variety of general chemistry analyzers
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration1.030 μg/mL
DescriptionGravity-Detect™
Quantity1 x 25 mL
TypeControl
Detectable AnalytesSpecific Gravity
Sample TypeUrine
Unit SizeEach
Contents & Storage
Room Temperature