1,3-Cyclopentanedione, 99%
1,3-Cyclopentanedione, 99%
1,3-Cyclopentanedione, 99%
Thermo Scientific Chemicals

1,3-Cyclopentanedione, 99%

CAS: 3859-41-4 | C5H6O2 | 98.10 g/mol
Have Questions?
Catalog NumberQuantityPackaging
17572025025 gGlass bottle
Catalog number 175720250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3859-41-4
IUPAC Namecyclopentane-1,3-dione
Molecular FormulaC5H6O2
InChI KeyLOGSONSNCYTHPS-UHFFFAOYSA-N
SMILESO=C1CCC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Appearance (Form)Crystalline powder
Melting point148°C to 153°C
Appearance (Color)Ochre to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only