1,1,4,4-Tetraphenyl-1,3-butadiene, 99%
1,1,4,4-Tetraphenyl-1,3-butadiene, 99%
1,1,4,4-Tetraphenyl-1,3-butadiene, 99%
Thermo Scientific Chemicals

1,1,4,4-Tetraphenyl-1,3-butadiene, 99%

CAS: 1450-63-1 | C28H22 | 358.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 176540250
Price (USD)
504.65
Special offer
Online exclusive
Ends: 28-Dec-2026
594.00
Save 89.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
504.65
Special offer
Online exclusive
Ends: 28-Dec-2026
594.00
Save 89.35
Each
Chemical Identifiers
CAS1450-63-1
IUPAC Name(1,4,4-triphenylbuta-1,3-dien-1-yl)benzene
Molecular FormulaC28H22
InChI KeyKLCLIOISYBHYDZ-UHFFFAOYSA-N
SMILESC(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or needles
Melting point195°C to 200°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only