Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water
Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water
Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water
Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water
Thermo Scientific Chemicals

Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water

CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 176610010
Price (USD)
621.65
Special offer
Online exclusive
Ends: 28-Dec-2026
731.00
Save 109.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
621.65
Special offer
Online exclusive
Ends: 28-Dec-2026
731.00
Save 109.35
Each
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Color scale=<150 APHA
Titration with HCl38 to 42 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Phase transfer catalyst
RUO – Research Use Only