Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
Thermo Scientific Chemicals

Tetrahydrofuran, 99+%, extra pure, stabilized with BHT

CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 176630010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Water=<0.05 % (Coulometric)
Peroxides=<0.0025 % (as H2O2)
Residue after evaporation=<0.005 %
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only