Borane-N,N-diethylaniline complex, 97%
Borane-N,N-diethylaniline complex, 97%
Borane-N,N-diethylaniline complex, 97%
Thermo Scientific Chemicals

Borane-N,N-diethylaniline complex, 97%

CAS: 13289-97-9 | C10H15BN | 160.05 g/mol
Quantity:
25 g
100 g
250 g
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 176771000
Price (JPY)
-
Quantity:
100 g
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13289-97-9
IUPAC NameN,N-diethylaniline boron
Molecular FormulaC10H15BN
InChI KeyKHYAFFAGZNCWPT-UHFFFAOYSA-N
SMILES[B].CCN(CC)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration>=96.0 % (Active hydrogen)
Refractive index1.5220 to 1.5250 (20°C, 589 nm)
Appearance (Color)Clear colorless to amber
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only