tert-Butyl methyl ether, 99%, pure
tert-Butyl methyl ether, 99%, pure
tert-Butyl methyl ether, 99%, pure
Thermo Scientific Chemicals

tert-Butyl methyl ether, 99%, pure

CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
25 L
Packaging:
View all
Plastic Drum
Catalog number 177040100
Price (USD)
335.65
Special offer
Online exclusive
Ends: 28-Dec-2026
395.00
Save 59.35
Each
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
335.65
Special offer
Online exclusive
Ends: 28-Dec-2026
395.00
Save 59.35
Each
Chemical Identifiers
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=98.5 %
Infrared spectrumConforms
Water=<0.01 % (Coulometric)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only