Borane dimethyl sulfide complex, 94%, AcroSeal™
Borane dimethyl sulfide complex, 94%, AcroSeal™
Borane dimethyl sulfide complex, 94%, AcroSeal™
Thermo Scientific Chemicals

Borane dimethyl sulfide complex, 94%, AcroSeal™

CAS: 13292-87-0 | C2H9BS | 75.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 177061000
Price (USD)
140.65
Online exclusive
156.00
Save 15.35
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
140.65
Online exclusive
156.00
Save 15.35
Each
Chemical Identifiers
CAS13292-87-0
IUPAC Name(methylsulfanyl)methane borane
Molecular FormulaC2H9BS
InChI KeyRMHDLBZYPISZOI-UHFFFAOYSA-N
SMILESB.CSC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to amber
Appearance (Form)Solution
Strength>=92 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only