Borane-dimethylamine complex, 98+%
Borane-dimethylamine complex, 98+%
Borane-dimethylamine complex, 98+%
Thermo Scientific Chemicals

Borane-dimethylamine complex, 98+%

CAS: 74-94-2 | C2H10BN | 58.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 177310050
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.60
Save 5.95
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.60
Save 5.95
Each
Chemical Identifiers
CAS74-94-2
IUPAC Namedimethylamine borane
Molecular FormulaC2H10BN
InChI KeyFTDUHBOCJSQEKS-UHFFFAOYSA-N
SMILESB.CNC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granules or crystals
Melting point33°C to 37°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration with Na2S2O3>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only