N-tert-Butyl-alpha-phenylnitrone, 98%
N-tert-Butyl-alpha-phenylnitrone, 98%
N-tert-Butyl-alpha-phenylnitrone, 98%
Thermo Scientific Chemicals

N-tert-Butyl-alpha-phenylnitrone, 98%

CAS: 3376-24-7 | C11H15NO | 177.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Packaging:
Glass bottle
Catalog number 177350250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3376-24-7
IUPAC NameN-tert-butyl-1-phenylmethanimine oxide
Molecular FormulaC11H15NO
InChI KeyIYSYLWYGCWTJSG-UHFFFAOYSA-N
SMILESCC(C)(C)[N+]([O-])=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point71°C to 75°C
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Appearance (Color)White to light beige
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only