Poly(acrylic acid), sec. stand., aver. MW 1,080,000, aver. MN 135,000
Poly(acrylic acid), sec. stand., aver. MW 1,080,000, aver. MN 135,000
Poly(acrylic acid), sec. stand., aver. MW 1,080,000, aver. MN 135,000
Thermo Scientific Chemicals

Poly(acrylic acid), sec. stand., aver. MW 1,080,000, aver. MN 135,000

CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
17806010010 gGlass bottle
Catalog number 178060100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9003-01-4
IUPAC NamePoly(acrylic acid)
Molecular Formula(C3H4O2)n
SMILESOC(=O)C(-*)C-*
MDL NumberMFCD00084394
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<10 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only