Polyethyleneimine, approx. M.N. 60,000, 50 wt% aq. solution, branched
Polyethyleneimine, approx. M.N. 60,000, 50 wt% aq. solution, branched
Polyethyleneimine, approx. M.N. 60,000, 50 wt% aq. solution, branched
Thermo Scientific Chemicals

Polyethyleneimine, approx. M.N. 60,000, 50 wt% aq. solution, branched

CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 178572500
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9002-98-6
Molecular Formula((C2H5N)zC2H4N)y(C2H5N)x
SMILES*-CCNCCN(-*)CCN-*
MDL NumberMFCD00084427
SpecificationsSpecification SheetSpecification Sheet
Assay48 to 52 % Solids
Appearance (Form)Clear viscous liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only