Poly(ethylene oxide), M.W. 100,000 to 200,000
Poly(ethylene oxide), M.W. 100,000 to 200,000
Poly(ethylene oxide), M.W. 100,000 to 200,000
Thermo Scientific Chemicals

Poly(ethylene oxide), M.W. 100,000 to 200,000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
Catalog number 178582500
Price (USD)
135.00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
135.00
Each
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Viscosity80 to 200 mPa.s (10 % w/w in water at 25°C)
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<2 % (10 g, 50°C, 5 h, 10 mmHg)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only