Coming soon: We’ll be introducing a new equivalent SKU for this product with updated branding. There will be no change to the formulation, performance, or pack configuration.
Ammonium Persulfate
Thermo Scientific™

Ammonium Persulfate

Thermo Scientific Pierce Ammonium Persulfate (APS) is an oxidizing agent that is used with TEMED to catalyze the polymerization ofRead more
Have Questions?
Catalog NumberQuantity
178744 x 25 g
Catalog number 17874
Price (USD)
72.25
Each
Quantity:
4 x 25 g
Request bulk or custom format
Price (USD)
72.25
Each
Thermo Scientific Pierce Ammonium Persulfate (APS) is an oxidizing agent that is used with TEMED to catalyze the polymerization of acrylamide and bisacrylamide to prepare polyacrylamide gels for electrophoresis.

Ammonium persulfate (APS) is an oxidizing agent that is often used with tetramethylethylenediamine (TEMED, Part No. 17919) to catalyze the polymerization of acrylamide and bisacrylamide to prepare polyacrylamide gels for electrophoresis.

Details:

• Name: Ammonium persulfate (APS)
• Formula: (NH4)2S2O8
• CAS number: 7727-54-0
• Molecular weight: 228.2
• Purity: ≥98.0%
• Insolubles: ≤0.005%
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormPowder
Quantity4 x 25 g
Product TypeAmmonium Persulfate
Unit SizeEach
Contents & Storage
Store below 23°C.

Frequently asked questions (FAQs)

Do you carry ammonium persulfate and TEMED?

Yes, details below:

- Ammonium Persulfate: Cat. No. 17874
- TEMED: Cat. No. 15524010 and Cat. No. 17919

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.