Do you have a protocol for ethidium bromide staining in agarose gels?
For use in agarose gels:
- Add ethidium bromide to melted agarose to a final concentration of 0.5 µg/mL. Do not melt agarose that already contains ethidium bromide.
For staining agarose gels after electrophoresis:
- You can stain gels that have been run in the absence of ethidium bromide by covering the gel in 0.5 µg/mL ethidium bromide in water and gently agitating for 10 to 30 minutes.
- If necessary, gels can be destained by shaking in H2O for an additional 30 minutes. To stain RNA gels, you should minimize staining time and definitely include a destaining period.
- Add ethidium bromide to melted agarose to a final concentration of 0.5 µg/mL. Do not melt agarose that already contains ethidium bromide.
For staining agarose gels after electrophoresis:
- You can stain gels that have been run in the absence of ethidium bromide by covering the gel in 0.5 µg/mL ethidium bromide in water and gently agitating for 10 to 30 minutes.
- If necessary, gels can be destained by shaking in H2O for an additional 30 minutes. To stain RNA gels, you should minimize staining time and definitely include a destaining period.