Phenol, Saturated - FAQs
Phenol, Saturated - FAQs
View additional product information for Phenol, Saturated - FAQs (17914)
1 product FAQs found
What buffer is supplied with Phenol, Saturated (Cat. No. 17914)?
Phenol, Saturated (Cat. No. 17914) is supplied with Tris buffer.