1-Methylisatin, 98%
1-Methylisatin, 98%
1-Methylisatin, 98%
Thermo Scientific Chemicals

1-Methylisatin, 98%

CAS: 2058-74-4 | C9H7NO2 | 161.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1793500101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 179350010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2058-74-4
IUPAC Name1-methyl-2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC9H7NO2
InChI KeyVCYBVWFTGAZHGH-UHFFFAOYSA-N
SMILESCN1C(=O)C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point129°C to 133°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only