Nunc™ Cell Scrapers
Thermo Scientific™

Nunc™ Cell Scrapers

Conduct a variety of applications with sterile, nonpyrogenic Thermo Scientific™ Nunc™ Cell Scrapers, which feature an adjustable blade.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric) Handle
179707PK32 cm
17969323 cm
179693PK23 cm
17970732 cm
Catalog number 179707PK
Price (USD)
308.65
Online exclusive
336.00
Save 27.35
Pack of 50
Length (Metric) Handle:
32 cm
Price (USD)
308.65
Online exclusive
336.00
Save 27.35
Pack of 50
Featuring an adjustable blade, the sterile, nonpyrogenic Thermo Scientific™ Nunc™ Cell Scraper allows for convenient mechanical harvesting of the adherent cells from culture vessels. Comes in 2 lengths to accommodate wide range of culture wares.
Features:
  • Flexible polystyrene handles
  • Sterilized and individually wrapped

Ordering Information:

  • 179707: for use with 75 to 175cm2 flasks
  • 179693: for use with 25 to 80cm2 flasks
Warranty: 90 days.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)75 to 175 cm&Sup2; Flask
Length (English) Handle12.5 in.
Length (Metric) Handle32 cm
No. per Case250/Cs.
No. per Pack1/Pk.
SterilitySterile
Length (English) Blade0.6299 in.
Length (Metric) Blade17.5 mm
MaterialPolystyrene (Handle)
Product TypeCell Scraper
Width (English) Blade0.6889 in.
Width (Metric) Blade17.5 mm
Unit SizePack of 50

Frequently asked questions (FAQs)

Can I use a Nunc Cell Scraper with a Nunc TripleFlask that has Nunclon Delta surface treatment?

The design of the Nunc TripleFlask does not allow for mechanical scraping. However, cells are easily harvested using protein digesting enzymes, such as trypsin, collagenase, and Pronase or by using non-enzymatic dissociation solutions.
The following is a document link regarding culturing techniques for Nunc TripleFlask:
https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/culturing-technique-nunc-tripleflask-technical-note.pdf

Are your Nunc Cell Scrapers flexible or rigid? What material are they made of?

Our Nunc Cell Scraper blade is rigid and made from polyethylene. It is hinged to allow for positioning at different angles for scraping cells using the windshield wiper or hoe method.