2-Bromonaphthalene, 99%
2-Bromonaphthalene, 99%
2-Bromonaphthalene, 99%
Thermo Scientific Chemicals

2-Bromonaphthalene, 99%

CAS: 580-13-2 | C10H7Br | 207.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 179710050
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.80
Save 7.15
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.80
Save 7.15
Each
Chemical Identifiers
CAS580-13-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light beige
Appearance (Form)Crystalline powder and/or chunks
GC>=98.5 %
Infrared spectrumConforms
Melting point53°C to 57°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only