2'-Bromoacetophenone, 98+%
2'-Bromoacetophenone, 98+%
2'-Bromoacetophenone, 98+%
Thermo Scientific Chemicals

2'-Bromoacetophenone, 98+%

CAS: 2142-69-0 | C8H7BrO | 199.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 179760050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2142-69-0
IUPAC Name1-(2-bromophenyl)ethan-1-one
Molecular FormulaC8H7BrO
InChI KeyPIMNFNXBTGPCIL-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to orange
GC>=98.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5650 to 1.5690 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only