Product Image
Invitrogen™

eBioscience™ Avidin HRP

The avidin horseradish peroxidase is commonly used as a second step for detection of biotinylated antibodies by ELISA and westernRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
18-4100-511 mL
18-4100-94500 μL
Catalog number 18-4100-51
Price (USD)
181.65
Online exclusive
187.00
Save 5.35
Each
Quantity:
1 mL
Price (USD)
181.65
Online exclusive
187.00
Save 5.35
Each
The avidin horseradish peroxidase is commonly used as a second step for detection of biotinylated antibodies by ELISA and western blot. Avidin binds to biotin with high affinity. [Note: Sodium azide is an inhibitor of horseradish peroxidase enzymatic activity and should not be added to the AV-HRP diluent.]

Conjugate
HRP

Reported Application
Western Blotting, ELISA

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeHRP (Horseradish Peroxidase)
Quantity1 mL
Target MoleculeBiotin
FormLiquid
Product LineeBioscience
Unit SizeEach

Frequently asked questions (FAQs)

What other reagents will I need to run an ELISA using the antibody pairs?

When working with our matched pairs or uncoated ELISA kits, please refer to the user guide provided with your antibody-matched pair for kit-specific guidance. When designing your own ELISA, please utilize our ELISA Builder tool for reagent selection as well as our ELISA Development and Optimization pages for futher guidance.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.