1-Adamantanemethanol, 99%
1-Adamantanemethanol, 99%
1-Adamantanemethanol, 99%
Thermo Scientific Chemicals

1-Adamantanemethanol, 99%

CAS: 770-71-8 | C11H18O | 166.26 g/mol
Have Questions?
Catalog NumberQuantityPackaging
18005010010 gGlass bottle
Catalog number 180050100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS770-71-8
IUPAC Name(adamantan-1-yl)methanol
Molecular FormulaC11H18O
InChI KeyMDVGOOIANLZFCP-UHFFFAOYSA-N
SMILESOCC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point115°C to 118°C
GC>=98.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only