Actin (Smooth Muscle) Mouse mAb (clone 1A4), concentrate - FAQs
Actin (Smooth Muscle) Mouse mAb (clone 1A4), concentrate - FAQs
View additional product information for Actin (Smooth Muscle) Mouse mAb (clone 1A4), concentrate - FAQs (180106)
1 product FAQs found
Do you offer an alternative to the discontinued Actin (Smooth Muscle) Mouse mAb (clone 1A4), concentrate (Cat. No. 180106)?
Unfortunately, we do not offer an alternative for this product.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.