LN2 Measuring Stick - FAQs
LN2 Measuring Stick - FAQs
View additional product information for LN2 Measuring Stick - FAQs (180143)
1 product FAQs found
How long is the LN2 Measuring Stick?
The LN2 measuring stick has a total length of ~114 cm (~45 inches) and the measuring portion of the stick is scaled to ~100 cm (40+ inch).