Naphthalene, 99+%, scintillation grade
Naphthalene, 99+%, scintillation grade
Naphthalene, 99+%, scintillation grade
Thermo Scientific Chemicals

Naphthalene, 99+%, scintillation grade

CAS: 91-20-3 | C10H8 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Catalog number 180200050
Price (USD)
35.65
Online exclusive
39.40
Save 3.75
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
35.65
Online exclusive
39.40
Save 3.75
Each
Chemical Identifiers
CAS91-20-3
IUPAC Namenaphthalene
Molecular FormulaC10H8
InChI KeyUFWIBTONFRDIAS-UHFFFAOYSA-N
SMILESC1=CC2=CC=CC=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Form)Powder or crystals or flakes
Infrared spectrumConforms
Solubility(5 % in methanol) Clear colorless
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For fluorescence/luminescence spectroscopy
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only