Orion™ 1803 Steam Sample Conditioner
Thermo Scientific™

Orion™ 1803 Steam Sample Conditioner

Take a high-pressure steam sample and reduce the temperature and pressure down to the levels required by our online analyzers.
Have Questions?
Catalog NumberTemperature (Metric) Max
180300538°C
Catalog number 180300
Price (USD)
13,590.00
Online exclusive
15,100.00
Save 1,510.00
Each
Temperature (Metric) Max:
538°C
Price (USD)
13,590.00
Online exclusive
15,100.00
Save 1,510.00
Each

Take high-pressure steam samples at the reduced temperature and pressure levels required by our online analyzers using the Thermo Scientific™ Orion™ 1803 Steam Sample Conditioner. Process steam is used in many industries; it is the analysis of this steam that has made Orion online analyzers so important in the power, paper, petrochemical and other industries.

  • Handles pressures of up to 20897KPa (3030psi)— average pressure of process steam is approximately 6896KPa (1000psi)
  • Reduces the temperature of the sample from 538°C (1000°F) to under 38°C (100°F)
  • Requires no electrical hook up, so it can be used in multiple countries regardless of the voltage requirements
  • Can also be used in hazardous locations (e.g. petrochem) without any danger of explosion
  • Has a mechanical temperature shut off valve which will trip if the temperature exceeds 40°C (104°F)
  • Avoids costly damage if cooler fails or the cooling water becomes too hot; valve diverts the hot sample away from online monitor
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Depth (English)6.5 in.
Depth (Metric)17 cm
Description1803 Steam Sample Conditioner
Height (English)36 in.
Height (Metric)91.5 cm
Temperature (English) Operating1000°F Max.
Temperature (Metric) Max538°C
Width (English)12 in.
Width (Metric)30.5 cm
TypeSteam Sample Conditioner
Unit SizeEach