Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000
Thermo Scientific Chemicals

Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Have Questions?
Catalog NumberQuantity
1803000101 kg
Catalog number 180300010
Price (EUR)
199,00
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
199,00
Each
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Granular powder
Infrared spectrumConforms
Volatile matter=<5 %
Ash=<1 % (as Na2O)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only