tert-Butyl hydroperoxide, 70% Solution in water
tert-Butyl hydroperoxide, 70% Solution in water
tert-Butyl hydroperoxide, 70% Solution in water
Thermo Scientific Chemicals

tert-Butyl hydroperoxide, 70% Solution in water

CAS: 75-91-2 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 180345000
Price (EUR)
46,65
Online exclusive
52,00
Save 5,35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
46,65
Online exclusive
52,00
Save 5,35 (10%)
Each
Chemical Identifiers
CAS75-91-2
IUPAC Name2-methylpropane-2-peroxol
Molecular FormulaC4H10O2
InChI KeyCIHOLLKRGTVIJN-UHFFFAOYSA-N
SMILESCC(C)(C)OO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Appearance (Form)Clear solution
Titration Iodometric68.5 to 71.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only