Product Image

Tryptose Phosphate Broth

Gibco™ Tryptose Phosphate Broth is a cell culture supplement for BHK-21 cells used for vaccine production. Tryptose Phosphate Broth containsRead more
Have Questions?
Catalog NumberQuantity
18050039100 mL
Catalog number 18050039
Price (BRL)
191,16
Each
Quantity:
100 mL
Price (BRL)
191,16
Each
Gibco™ Tryptose Phosphate Broth is a cell culture supplement for BHK-21 cells used for vaccine production. Tryptose Phosphate Broth contains tryptose (an enzymatic hydrolysis of protein), dextrose, sodium chloride and disodium phosphate.

Product Use
For Research Use Only. Not for use in diagnostic procedures.

cGMP Manufacturing and Quality System
Gibco™ Tryptose Phosphate Broth is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Quantity100 mL
Shelf Life12 Months
Shipping ConditionRoom Temperature
Product TypeTryptose Phosphate Broth
Unit SizeEach
Contents & Storage
Storage conditions: 2 – 8°C
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

What is the concentration of tryptose phosphate solution in Tryptose Phosphate Broth (Cat. No. 18050039)?

The Tryptose Phosphate Broth contains 29.5 g/L tryptose phosphate.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.