Isopropyl ether, 99+%, extra pure, stabilized with BHT
Isopropyl ether, 99+%, extra pure, stabilized with BHT
Isopropyl ether, 99+%, extra pure, stabilized with BHT
Thermo Scientific Chemicals

Isopropyl ether, 99+%, extra pure, stabilized with BHT

CAS: 108-20-3 | C6H14O | 102.177 g/mol
数量:
1 L
2.5 L
25 L
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 180680025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS108-20-3
IUPAC Name2-(propan-2-yloxy)propane
Molecular FormulaC6H14O
InChI KeyZAFNJMIOTHYJRJ-UHFFFAOYSA-N
SMILESCC(C)OC(C)C
さらに表示
Color scale=<10 APHA
Refractive index1.3670 to 1.3690 (20°C, 589 nm)
GC>=99.0 %
Residue after evaporation=<0.01 %
Peroxides=<0.002 % (as H2O2)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only