Isopropyl acetate, 99+%, pure
Isopropyl acetate, 99+%, pure
Isopropyl acetate, 99+%, pure
Thermo Scientific Chemicals

Isopropyl acetate, 99+%, pure

CAS: 108-21-4 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Catalog number 180800250
Price (USD)
807.65
Special offer
Online exclusive
Ends: 28-Dec-2026
950.00
Save 142.35
Each
Quantity:
25 L
Request bulk or custom format
Price (USD)
807.65
Special offer
Online exclusive
Ends: 28-Dec-2026
950.00
Save 142.35
Each
Chemical Identifiers
CAS108-21-4
IUPAC Namepropan-2-yl acetate
Molecular FormulaC5H10O2
InChI KeyJMMWKPVZQRWMSS-UHFFFAOYSA-N
SMILESCC(C)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only