SuperScript™ IV RT Reaction Buffer - FAQs
SuperScript™ IV RT Reaction Buffer - FAQs
View additional product information for SuperScript™ IV RT Reaction Buffer - FAQs (18090050B)
1 product FAQs found
Can I use the SuperScript IV RT Reaction Buffer with SuperScript II RT or SuperScript III RT enzymes?
No, we do not recommend using SuperScript IV RT Reaction Buffer with the SuperScript II RT or SuperScript III RT enzymes. For optimal performance, we recommended using the matching reaction buffer for each enzyme.