SuperScript™ IV First-Strand Synthesis System with ezDNase™ Enzyme, Each - FAQs
SuperScript™ IV First-Strand Synthesis System with ezDNase™ Enzyme, Each - FAQs
View additional product information for SuperScript™ IV First-Strand Synthesis System - FAQs (18091150, 18091050, 18091200, 18091300)
1 product FAQs found
Can I use SuperScript IV First-Strand Synthesis System with ezDNase Enzyme (Cat. No. 18091150, 18091300) for qPCR with a TaqMan assay?
While the SuperScript IV First-Strand Synthesis System with ezDNase Enzyme is compatible with TaqMan assays, for RT-qPCR applications, we recommend using the SuperScript IV VILO Master Mix. The cDNA synthesis reaction setup with this master mix requires fewer pipetting steps and therefore reduces variation in the data.