BioPrime™ Array CGH Genomic Labeling System - FAQs
BioPrime™ Array CGH Genomic Labeling System - FAQs
View additional product information for BioPrime™ Array CGH Genomic Labeling System - FAQs (18095011)
2 product FAQs found
Can the PureLink columns provided to purify labeled cDNA probes be purchased separately? Which stand-alone columns are compatible?
The PureLink PCR purification system (Cat. Nos. K3100-01, K3100-02) can be used to purify labeled probe. Because tubes provided with stand-alone kit are not amber, be sure to protect your probe from light if you are doing direct labeling. Please contact Technical Support for the protocol.
Can the BioPrime Array CGH Genomic Labeling System be used for SNP analysis?
We do not recommend using the BioPrime Array CGH Genomic Labeling System to amplify genomic DNA for SNP analysis because the exo-Klenow polymerase has no proofreading activity.