1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene
1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene
1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene
Thermo Scientific Chemicals

1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene

CAS: 90-13-1 | C10H7Cl | 162.62 g/mol
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 180970025
Price (JPY)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS90-13-1
IUPAC Name1-chloronaphthalene
Molecular FormulaC10H7Cl
InChI KeyJTPNRXUCIXHOKM-UHFFFAOYSA-N
SMILESClC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
AppearanceMay darken on storage
Infrared spectrumConforms
HPLC>=80 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only