Cumene, 99%, pure
Cumene, 99%, pure
Cumene, 99%, pure
Thermo Scientific Chemicals

Cumene, 99%, pure

CAS: 98-82-8 | C9H12 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 181010010
Price (USD)
49.65
Online exclusive
55.30
Save 5.65
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
49.65
Online exclusive
55.30
Save 5.65
Each
Chemical Identifiers
CAS98-82-8
IUPAC Name(propan-2-yl)benzene
Molecular FormulaC9H12
InChI KeyRWGFKTVRMDUZSP-UHFFFAOYSA-N
SMILESCC(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4900 to 1.4930 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only