NTP Mix (10 mM)
Invitrogen™

NTP Mix (10 mM)

ATP, CTP, GTP, UTP (nucleoside 5´-triphosphates), and 10 mM NTP Mix are suitable for use in in vitro transcription reactionsRead more
Have Questions?
Catalog NumberQuantity
1810901720 μmol
Catalog number 18109017
Price (USD)
300.65
Online exclusive
327.00
Save 26.35
Each
Quantity:
20 μmol
Request bulk or custom format
Price (USD)
300.65
Online exclusive
327.00
Save 26.35
Each

ATP, CTP, GTP, UTP (nucleoside 5´-triphosphates), and 10 mM NTP Mix are suitable for use in in vitro transcription reactions with SP6, T3, and T7 RNA polymerases for production of RNA probes, or RNA transcripts for translation in in vitro translation extracts. These nucleotides are supplied as convenient, ready-to-use solutions at a concentration of 10 mM in 1 mM Tris-HCl (pH 7.5).

Related products:
• 10 mM ATP
• 10 mM CTP
• 10 mM GTP
• 10 mM UTP

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)in vitro Transcription
FormLiquid
Label or DyeUnlabeled
Quantity20 μmol
Shipping ConditionApproved for shipment on Wet or Dry Ice
Concentration10 mM
Unit SizeEach
Contents & Storage
NTP Mix, 10 mM (500 μL)

Store at –20°C.