n-Butyllithium, 1.6M solution in hexanes, AcroSeal™
n-Butyllithium, 1.6M solution in hexanes, AcroSeal™
n-Butyllithium, 1.6M solution in hexanes, AcroSeal™
Thermo Scientific Chemicals

n-Butyllithium, 1.6M solution in hexanes, AcroSeal™

CAS: 109-72-8 | C4H9Li | 64.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 181278000
Price (USD)
171.65
Online exclusive
190.00
Save 18.35
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
171.65
Online exclusive
190.00
Save 18.35
Each
Chemical Identifiers
CAS109-72-8
IUPAC Namebutyllithium
Molecular FormulaC4H9Li
InChI KeyMZRVEZGGRBJDDB-UHFFFAOYSA-N
SMILES[Li]CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Free base=<1 %
Appearance (Color)Clear to hazy colorless to yellow
AppearanceMay develop some turbidity or precipitate
Titration Active base14.0 to 16.0 %
Titration Active base1.49 to 1.70 M
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only