Methyl acetate, 99%, extra pure
Methyl acetate, 99%, extra pure
Methyl acetate, 99%, extra pure
Thermo Scientific Chemicals

Methyl acetate, 99%, extra pure

CAS: 79-20-9 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 181380010
Price (USD)
92.20
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
92.20
Each
Chemical Identifiers
CAS79-20-9
IUPAC Namemethyl acetate
Molecular FormulaC3H6O2
InChI KeyKXKVLQRXCPHEJC-UHFFFAOYSA-N
SMILESCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=98.5 %
Refractive index1.3600 to 1.3630 (20°C, 589 nm)
Water=<0.1 % (K.F.)
Residue after evaporation=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only