4-Bromobenzyl alcohol, 99%
4-Bromobenzyl alcohol, 99%
4-Bromobenzyl alcohol, 99%
Thermo Scientific Chemicals

4-Bromobenzyl alcohol, 99%

CAS: 873-75-6 | C7H7BrO | 187.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 181770100
Price (USD)
56.65
Online exclusive
63.10
Save 6.45
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
56.65
Online exclusive
63.10
Save 6.45
Each
Chemical Identifiers
CAS873-75-6
IUPAC Name(4-bromophenyl)methanol
Molecular FormulaC7H7BrO
InChI KeyVEDDBHYQWFOITD-UHFFFAOYSA-N
SMILESOCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Melting point75°C to 78°C
Appearance (Form)Crystalline powder or crystals
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only