9-Anthracenemethanol, 98%
9-Anthracenemethanol, 98%
9-Anthracenemethanol, 98%
Thermo Scientific Chemicals

9-Anthracenemethanol, 98%

CAS: 1468-95-7 | C15H12O | 208.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 181870250
Price (USD)
175.65
Online exclusive
195.00
Save 19.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
175.65
Online exclusive
195.00
Save 19.35
Each
Chemical Identifiers
CAS1468-95-7
IUPAC Name(anthracen-9-yl)methanol
Molecular FormulaC15H12O
InChI KeyJCJNNHDZTLRSGN-UHFFFAOYSA-N
SMILESOCC1=C2C=CC=CC2=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point160°C to 164°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only