Orion™ 2100 Series and 1800 Series Analyzer Reagents for Orion™ Online Monitors
Use these analyzer reagents for our process analyzers.
Have Questions?
Change view
Catalog Number
dimType
182011
181030
181130
181811
211190
211191
6 Options
Catalog number 182011
Price (USD)
219.60
Online exclusive
244.00
Save 24.40
Each
Price (USD)
219.60
Online exclusive
244.00
Save 24.40
Each
Use Thermo Scientific™ Orion™ 2100 Series and 1800 Series Analyzer Reagents for Orion™ Online Monitors with Orion™ process analyzers.
Reagents for the Orion 2109XP, 2110XP, 2111LL, 2111XP, 2117XP, 2118XP, 2120XP, 1809, 1810, 1811AO, 1811EL, 1817HL, 1817LL, 1818 and 1820 analyzers.
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)2109XP Fluoride, 2117XP Chloride and 2120XP Calcium Hardness Analyzers