Search
View additional product information for SuperScript™ Plasmid System for cDNA Synthesis and Plasmid Cloning with Gateway™ Technology - FAQs (18248013)
46 product FAQs found
Small-Scale Preparation of ss Phagemid DNA:
1. Pick a single colony of cells containing phagemid DNA and resuspend in 2 mL TBG (1.2% tryptone, 2.4% yeast extract 0.4% glycerol, 17 mM KH2PO4 and 55 mM KH2PO4 and 20 mM glucose) containing 100 µg/mL ampicillin in a 15 mL tube.
2. Immediately add 10 µL M13KO7 helper phage stock at 10E11 pfu/mL.
3. Incubate cells at 37 degrees C with vigorous agitation (275 rpm) for 2 hrs.
4. Add kanamycin to a final concentration of 75 µg/mL and incubate cells at 37 degrees C with vigorous agitation (275 rpm) for 18 to 24 hrs.
5. Transfer 1.5 mL of culture to a sterile microcentrifuge tube and pellet cells by centrifuging at 14,000 x g for 10 min at 4 degrees C.
6. Transfer supernatant to fresh microcentrifuge tube and repeat the centrifugation.
7. Transfer 1.2 mL supernatant to a fresh microcentrifuge tube and add 300 µL of 2.5 M NaCl in 40% PEG 4000.
8. Vortex and incubate on ice for 15 min.
9. Centrifuge at 14,000 x g for 15 min at 4 degrees C.
10. Resuspend the pellet in 50 µL TE and phenol extract to remove the viral coat.
11. Use 10 µL of the final 50 µL volume for gel analysis.
This protocol yields 0.5 to 1 µg ss phagemid DNA.
Large-Scale Preparation of ss Phagemid DNA:
1. Resuspend a single colony in 5 mL of TB or LB broth containing 100 µg/mL ampicillin in a 15-mL tube.
2. Shake at 37 degrees C and 275 rpm overnight.
3. Add 100 µL of the overnight culture to 200 mL LB broth and 100 µg/mL ampicillin in a 1 L flask. Incubate at 37 degrees C with shaking (275 rpm) for 3 hrs.
4. Add 200 µL of M13KO7 helper phage (1 x 10E11 pfu/mL) to the culture and continue to incubate for 2 hrs.
5. Add 1.5 mL of 1% (w/v) kanamycin to the cells for a final concentration of 75 µg/mL. Incubate the infected cells for an additional 18 to 24 h at 37 degrees C.
6. Centrifuge this culture at 16,000 x g for 15 min at 4 degrees C .
7. Filter the supernatant through a 0.2 µm sterile filter into an autoclaved centrifuge bottle. Add 40 µL of DNase I (50 units/µL) and incubate at room temperature for 3 hrs. This step should remove any residual ds plasmid DNA contamination.
8. Transfer 100 mL of the supernatant to another centrifuge bottle. Add 25 mL of 2.5 M NaCl in 40% PEG 4000 to each of the centrifuge bottles containing the supernatant.
9. Vortex the mixture, incubate on ice for 1 h, and centrifuge at 16,000 x g for 20 min at 4 degrees C.
10. Carefully discard the supernatant. To fully drain off the remaining solution from the pellets, place the bottles on an angle, with the pellet side facing up for 10 to 15 min. Remove the solution with a sterile Pasteur pipet.
11. Resuspend the pellets in 2 mL of TE buffer. Add 10 µL of proteinase K solution (20 mg/mL), 20 µL of 10% SDS, and incubate this mixture at 45 degrees C for 1 hr.
12. Transfer the digested mixture to three microcentrifuge tubes and extract four times with an equal volume of phenol:chloroform: isoamyl alcohol (25:24:1), precipitate with ethanol, and dissolve in 100 µL TE buffer.
13. Freeze the solubilized DNA at -20 degrees C for 1 hr and centrifuge in a microcentrifuge at 14,000 x g for 15 min at 4 degrees C.
14. Transfer the supernatant containing the ss plasmid DNA to a fresh tube and discard the polysaccharide pellet. Store the ssDNA at 4 degrees C.
15. Determine the DNA concentration (OD260).
This protocol yields ~100 to 200 µg ss phagemid DNA.
(1) Vieira, J. and Messing, J. (1987) Methods in Enzymology 153, 3.
To make a glycerol stock of lawn cells:
1) Streak cells on an LB plate and incubate overnight at 37C.
2) Pick a single colony from the plate into 50 ml LB or S.O.B. medium and incubate the culture at 37C until the OD550 reaches 2.0.
3) Centrifuge the cells in a clinical centrifuge at 3,000 x g for 10 min at 4C.
4) Resuspend the pellet in 5 ml 60% S.O.B. and 40% glycerol.
5) Aliquot the cells and freeze them in a dry ice/ethanol bath. Store the cells at -70C.
LB Medium (per liter):
- 10 g Tryptone (SELECT Peptone 140)
- 5 g Yeast Extract (SELECT Yeast Extract)
- 10 g NaCl
Add Distilled Water to a final volume of 1 L. Stir to dissolve, and autoclave.
S.O.B. Medium (per liter):
- 20 g Tryptone (SELECT Peptone 140)
- 5 g Yeast Extract (SELECT Yeast Extract)
- 10 ml 1 M NaCl
- 2.5 ml 1 M KCl
- 10 ml 1 M MgCl2
- 10 ml 1 M MgSO4
Add tryptone, yeast extract, NaCl, and KCl to a flask or bottle, and add Distilled Water to a final volume of 1 L (~980 ml). Stir to dissolve, autoclave, and cool to room temperature. Then add Mg++ stock solutions (1 M MgCl2/6H2O and 1 M MgSO4/7H2O, filter sterilized). Mg++ solutions should not be added before autoclaving, as this will likely result in precipitation.
Alternatively, amplification of plasmid libraries may be done on selective agar plates, as described below. Libraries containing expression vector plasmids such as pSPORT or pCMV SPORT should be grown under noninduced conditions (no lactose, no IPTG). The lac repressor gene encoded on the pSPORT plasmid is sufficient to prevent expression of the recombinant genes cloned into the multiple cloning site. Repression of expression is necessary to ensure the amplification of clones whose gene products may be harmful to survival of the E. coli host.
Recommended protocol for Library Amplification on Plates:
1. After titering the original library, plate the cells at a density of ~1 x 10E4 CFU per 100 mm plate or ~0.5 x 10E5 CFU per 150 mm plate. Use selective media containing the appropriate antibiotic.
2. Grow at 37°C until a thick lawn is visible (~6 to 12 h).
3. Add 5 ml per 100 mm plate or 15 ml per 150 mm plate of S.O.C. medium or LB broth.
4. Gently coax the bacterial lawn into the broth with a cell scraper (or a glass slide).
5. Collect the suspension and repeat steps 3 and 4 once more. Pool the broths, then incubate at 37°C for no longer than 1 h with constant swirling.
6a. If the bacterial suspension is highly dense, add glycerol to a concentration of 15% to 50%; aliquot the cells into cryovials; and immediately store them at -70°C.
6b. If the bacterial solution is too dilute, centrifuge at 7,000 x g for 10 min and resuspend in a smaller volume of 15% to 50% glycerol/media and freeze.
7. For use, thaw the cryovials and plate dilutions onto selective media.
NOTE:
-The viability of E. coli stored at -70°C will not change substantially for years, unless thawed and refrozen.
When attempting this protocol, we observed very low efficiencies (~10 colonies/plate). So just be aware that while technically possible, going directly into an LR reaction from a TOPO reaction is very inefficient and will result in a very low colony number, if any at all.
To attach a C-terminal tag to your gene of interest, the insert must lack its stop codon, and be in the correct reading frame for compatibility with our C-terminal tagged destination vectors. Again, TOPO adapted Gateway Entry vectors will automatically put the insert into the correct reading frame. If you do not want the C-terminal tag to be expressed, simply include a stop codon at the end of the insert that is in frame with the initial ATG.
Generally, you need to choose a destination vector before you design and clone your insert into the Entry vector. This will determine whether you need to include an initiating ATG or stop codon with your insert.
There is a protocol in which all 4 primers mentioned above are in a single PCR reaction. You can find this protocol at in the following article: Quest vol. 1, Issue 2, 2004. https://www.thermofisher.com/us/en/home/references/newsletters-and-journals/quest-archive.reg.in.html. The best ratio of the first gene-specific and the second attB primers was 1:10.
2. Protein Interaction Mapping in C. elegans Using Proteins Involved in Vulval Development. Science Jan 7th 2000; 287(5450), 116-122 Davy, A. et al.
3. A protein-protein interaction map of the Caenorhabditis elegans 26S proteosome. EMBO Reports (2001) 2 (9), p. 821-828. Walhout, A.J.M. and Vidal, M. (2001).
4. High-throughput Yeast Two-Hybrid Assays for Large-Scale Protein Interaction mapping. Methods: A Companion to Methods in Enzymology 24(3), pp.297-306
5. Large Scale Analysis of Protein Complexes Gavin, AC et al. Functional Organization of the Yeast Proteome by Systematic Analysis of Protein Complexes. Nature Jan 10th 2002, 415, p. 141-147.
6. Systematic subcellular localisation of proteins Simpson, J.C., Wellenreuther, R., Poustka, A., Pepperkok, R. and Wiemann, S.
7. Systematic subcellular localization of novel proteins identified by large-scale cDNA sequencing. EMBO Reports (2000) 1(3), pp. 287-292.
8. Protein-over expression and crystallography Evdokimov, A.G., Anderson, D.E., Routzahn, K.M. & Waugh, D.S.
9. Overproduction, purification, crystallization and preliminary X-ray diffraction analysis of YopM, an essential virulence factor extruded by the plague bacterium Yersinia pestis. Acta Crystallography (2000) D56, 1676-1679.
10. Evdokimov, et al. Structure of the N-terminal domain of Yersinia pestis YopH at 2.0 A resolution. Acta Crystallographica D57, 793-799 (2001).
11. Lao, G. et al. Overexpression of Trehalose Synthase and Accumulation of Intracellular Trehalose in 293H and 293FTetR:Hyg Cells. Cryobiology 43(2):106-113 (2001).
12. High-throughput cloning and expression Albertha J. M. Walhout, Gary F. Temple, Michael A. Brasch, James L. Hartley, Monique A. Lorson, Sander Van Den Huevel, and Marc Vidal.
13. Gateway Recombinational Cloning: Application to the Cloning of Large Numbers of Open Reading Frames or ORFeomes. Methods in Enzymology, Vol. 328, 575-592.
14. Wiemann, S. et.al., Toward a Catalog of Human Genes and Proteins: Sequencing and Analysis of 500 Novel Complete Protein Coding Human cDNAs, Genome Research (March 2001) Vol. 11, Issue 3, pp.422-435
15. Reviewed in NATURE: Free Access to cDNA provides impetus to gene function work. 15 march 2001, p. 289. Generating directional cDNA libraries using recombination
16. Osamu Ohara and Gary F. Temple. Directional cDNA library construction assisted by the in vitro recombination reaction. Nucleic Acids Research 2001, Vol. 29, no. 4. RNA interference (RNAi)
17. Varsha Wesley, S. et al. Construct design for efficient, effective and highthroughput gene silencing in plants. The Plant Journal 27(6), 581-590 (2001). Generation of retroviral constructs
18. Loftus S K et al. Generation of RCAS vectors useful for functional genomic analyses. DNA Res 31;8(5):221 (2001).
19. James L. Hartley, Gary F. Temple and Michael A. Brasch. DNA Cloning Using In Vitro Site-Specific Recombination. Genome Research (2000) 10(11), pp. 1788-1795.
20. Reboul et al. Open-reading frame sequence tags (OSTs) support the existence of at least 17,300 genes in C. elegans. Nature Genetics 27(3):332-226 (2001).
21. Kneidinger, B. et al. Identification of two GDP-6-deoxy-D-lyxo-4-hexulose reductase synthesizing GDP-D-rhamnose in Aneurinibacillus thermoaerophilus L420-91T*. JBC 276(8) (2001).
The region within brackets is where the site is "cut" and replaced by the attB1-fragment sequence to make an attL1 site. The sequence GTACAAA is the overlap sequence present in all att1 sites and is always "cut" right before the first G.
The overlap sequence in attP2 sites is CTTGTAC and cut before C. This is attP2:
ACAGGTCACT AATACCATCT AAGTAGTTGA TTCATAGTGA CTGGATATGT TGTGTTTTAC AGTATTATGT AGTCTGTTTT TTATGCAAAA TCTAATTTAA TATATTGATA TTTATATCAT TTTACGTTTC TCGTTCAGCT TTCTTGTACA AAGTTGGCAT TATAAGAAAG CATTGCTTAT AATTTGTTG CAACGAACAG GTCACTATCA GTCAAAATAA AATCATTATT
So, attL1 (Entry Clone) should be:
A ATAATGATTT TATTTTGACT GATAGTGACC TGTTCGTTGC AACAAATTGA TGAGCAATGC TTTTTTATAA TGCCAACT TT G TAC AAA AAA GC[A GGC T]NN NNN
attL2 (Entry Clone) should be:
NNN N[AC C]CA GCT TT CTTGTACA AAGTTGGCAT TATAAGAAAG CATTGCTTAT CAATTTGTTG CAACGAACAG GTCACTATCA GTCAAAATAA AATCATTATT
The sequence in brackets comes from attB, and N is your gene-specific sequence.
Note: When creating an Entry Clone through the BP reaction and a PCR product, the vector backbone is not the same as Gateway Entry vectors. The backbone in the case of PCR BP cloning is pDONR201.
Effects on protein function: A researcher (Simpson et al. EMBO Reports 11(31):287-292, 2000) demonstrated that GFP fusions (N- terminal and C-terminal) localized to the proper intracellular compartment. The expression constructs were generated using Gateway cloning, so the recombinant protein contained the attB1 or attB2 amino acid sequence. The localization function of the cloned recombinant proteins was preserved.
Effects on expression: We have seen no effect of the attB sites on expression levels in E. coli, insect and mammalian cells. The gus gene was cloned into bacterial expression vectors (for native and N-terminal fusion protein expression) using standard cloning techniques and expressed in bacteria. Gus was also cloned into Gateway Destination vectors (for native and N-terminal fusion expression) and expressed. When protein expression is compared, there was no difference in the amount of protein produced. This demonstrates that for this particular case, the attB sites do not interfere with transcription or translation.
Effects on solubility: A researcher at the NCI has shown that Maltose Binding Protein fusions constructed with Gateway Cloning were soluble. The fusion proteins expressed had the attB amino acid sequence between the Maltose Binding Protein and the cloned protein. It is possible that some proteins containing the attB sequence could remain insoluble when expressed in E.coli.
Effects on folding: Two Hybrids screens show the same interacters identified with and without the attB sequence. Presumably correct protein folding would be required for protein-protein interactions to take place. It is possible that some proteins containing the attB sequence may not fold correctly.
The location of translation initiation sites, stop codons, or fusion tags for expression must be considered in your initial cloning design. For example, if your destination vector contains an N-terminal tag but does not have a C-terminal tag, the vector should already contain the appropriate translation start site but the stop codon should be included in your insert.
These enzymes are more stable than the original BP and LR Clonase and can be stored at -20 degrees C for 6 months.