1-Phenyl-2-propanol, 98%
1-Phenyl-2-propanol, 98%
1-Phenyl-2-propanol, 98%
Thermo Scientific Chemicals

1-Phenyl-2-propanol, 98%

CAS: 14898-87-4 | C9H12O | 136.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 183060050
Price (USD)
38.65
Online exclusive
42.90
Save 4.25
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
38.65
Online exclusive
42.90
Save 4.25
Each
Chemical Identifiers
CAS698-87-3
IUPAC Name1-phenylpropan-2-ol
Molecular FormulaC9H12O
InChI KeyWYTRYIUQUDTGSX-UHFFFAOYNA-N
SMILESCC(O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless to almost colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5200 to 1.5240 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only