M13KO7 Helper Phage - FAQs
M13KO7 Helper Phage - FAQs
View additional product information for M13KO7 Helper Phage - FAQs (18311019)
1 product FAQs found
Do you offer an alternative for M13KO7 Helper Phage (Cat. No. 18311019)?
We do not offer an alternative for M13KO7 Helper Phage.