3' RACE System for Rapid Amplification of cDNA Ends
3´ RACE System is suitable for rapid amplification of cDNA ends (RACE) (1-3) and anchored PCR between a defined pointRead more
Have Questions?
Catalog Number
Quantity
18373019
20 Reactions
Catalog number 18373019
Price (USD)
861.65
Online exclusive
898.00
Save 36.35
Each
Quantity:
20 Reactions
Price (USD)
861.65
Online exclusive
898.00
Save 36.35
Each
3´ RACE System is suitable for rapid amplification of cDNA ends (RACE) (1-3) and anchored PCR between a defined point within mRNA and the 3´ poly(A) end (Figure 1). The system is useful for the amplification of rare messages for which little sequence information is available, and for capturing the 3´ end information of mRNA. The 3´ RACE System:
• Can be used with the 5´ RACE System to clone full-length sequences • Uses the advanced capabilities of SuperScript™ II RT for greater first-strand cDNA yields • Includes Control RNA and primers to monitor the performance of the system • Provides detailed protocols in the manual • Converts up to 1 μg of total RNA preparation or 100 ng of control RNA into cDNA in one reaction
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)cDNA Libraries & Library Construction
Product TypecDNA library
Quantity20 Reactions
Unit SizeEach
Contents & Storage
3´ RACE System components are listed in Table 1. Store the system at -20°C. Guaranteed stable for 6 months when properly stored.
Frequently asked questions (FAQs)
Can I purchase the Adapter Primer (AP), Universal Amplification Primer (UAP), and Abridged Universal Amplification Primer (AUAP) from the 3‘ RACE System as stand-alone items?
The Adapter Primer (AP), Universal Amplification Primer (UAP), and Abridged Universal Amplification Primer (AUAP) have been discontinued as stand-alone items. Their sequences can be found on Page 4 of the manual.
I'm getting PCR products from my 5' RACE, but they are not full length. What should I do?
The GeneRacer method is designed to ensure that only full-length messages are ligated to the GeneRacer RNA Oligo and PCR amplified after cDNA synthesis. It is highly recommended that you clone your RACE products and analyze at least 10-12 colonies to ensure that you isolate the longest message. Many genes do not have only one set of transcription start sites but rather multiple transcription start sites spanning sometimes just a few or other times a hundred or even more bases. Cloning of the RACE products and analyzing multiple colonies ensues that you detect the diversity of the heterogeneous transcription start sites of your gene. It is also possible that you might obtain PCR products that may not represent the full-length message for your gene. PCR products that do not represent full-length message may be obtained because:
-RNA degradation after the CIP reaction creates new truncated substrates with a 5' phosphate for ligation to the GeneRacer RNA Oligo. Be sure to take precautions to ensure that the RNA is not degraded.
-CIP dephosphorylation was incomplete. Increase the amount of CIP in the reaction or decrease the amount of RNA.
-PCR yielded a PCR artifact and not true ligation product. Optimize your PCR using the suggestions described above.
I'm seeing RACE PCR artifacts in my GeneRacer experiment. What am I doing wrong?
RACE PCR artifacts or nonspecific PCR bands can result from one or more of the following:
-Nonspecific binding of GSPs to other cDNAs resulting in the amplification of unrelated products as well as desired products.
-Nonspecific binding of GeneRacer primers to cDNA resulting in PCR products with GeneRacer primer sequence on both ends of the PCR product.
-RNA degradation.
-Contamination of PCR tubes or reagents.
Note: Artifacts usually result from less than optimal PCR conditions and can be identified in negative control PCR.
I'm getting unexpected bands after electrophoretic analysis of my amplified RT-PCR products. Can you please offer some suggestions?
Please see the following causes and suggestions:
Contamination by genomic DNA or an unexpected splice variant - Pretreat RNA with DNase I, amplification grade (Cat. No 18068015). Design primers that anneal to sequences in exons on both sides of an intron or at the exon/exon boundary of the mRNA to differentiate between amplified cDNA and potential contaminating genomic DNA. To test if products were derived from DNA, perform a minus RT control.
Nonspecific annealing of primers - Vary the PCR annealing conditions. Use a hot-start PCR polymerase. Optimize magnesium concentration for each template and primer combination.
Primers formed dimers - Design primers without complementary sequences at the 3' ends.
I'm getting no bands after electrophoretic analysis of my amplified RT-PCR products. Can you please offer some tips?
Please see the following causes and suggestions:
Procedural error in first-strand cDNA synthesis - Use high-quality RNA as a control to verify the efficiency of the first-strand reaction.
RNase contamination - Add control RNA to sample to determine if RNase is present in the first-strand reaction. Use an RNase inhibitor in the first-strand reaction.
Polysaccharide co-precipitation of RNA - Precipitate RNA with lithium chloride to remove polysaccharides, as described in Sambrook et al.
Target mRNA contains strong transcriptional pauses - Use random hexamers instead of oligo(dT) in the first-strand reaction, increase the temperature, and use PCR primers closer to the 3' terminus of the target cDNA.
Too little first-strand product was used in PCR - Use up to 10% of first-strand reaction per 50 mL PCR.
Gene-specific primer was used for first-strand synthesis - Try another set of GSP or switch to oligo(dT). Make sure the GSP is the antisense of the sequence.
Inhibitors of RT present - Remove inhibitors by ethanol precipitation of mRNA preparation before the first-strand reaction. Include a 70% (v/v) ethanol wash of the mRNA pellet. Note: inhibitors of RT include SDS, EDTA, guanidinium salts, formamide, sodium pyrophosphate, and spermidine.
RNA has been damaged or degraded - Ensure that high-quality, intact RNA is being used.
Annealing temperature is too high - Decrease temperature as necessary and/or use touchdown PCR.
Recovery of an arenavirus entirely from RNA polymerase I/II-driven cDNA.
Authors:Flatz L,Bergthaler A,de la Torre JC,Pinschewer DD
Journal:Proceedings of the National Academy of Sciences of the United States of America
PubMed ID:16537369
The prototypic arenavirus lymphocytic choriomeningitis virus has been a primary workhorse of viral immunologists for almost a century, and it has served as an important model for studying basic principles of arenavirus molecular biology. Its negative-stranded bisegmented RNA genome has, however, posed a major obstacle to attempts at manipulating the ... More
Reconstitution of human DNA polymerase delta using recombinant baculoviruses: the p12 subunit potentiates DNA polymerizing activity of the four-subunit enzyme.
Authors: Podust Vladimir N; Chang Long-Sheng; Ott Robert; Dianov Grigory L; Fanning Ellen;
Journal:J Biol Chem
PubMed ID:11711545
'Eukaryotic DNA polymerase delta is thought to consist of three (budding yeast) or four subunits (fission yeast, mammals). Four human genes encoding polypeptides p125, p50, p66, and p12 have been assigned as subunits of DNA polymerase delta. However, rigorous purification of human or bovine DNA polymerase delta from natural sources ... More
The cathepsin B of Toxoplasma gondii, toxopain-1, is critical for parasite invasion and rhoptry protein processing.
Authors: Que Xuchu; Ngo Huân; Lawton Jeffrey; Gray Mary; Liu Qing; Engel Juan; Brinen Linda; Ghosh Partho; Joiner Keith A; Reed Sharon L;
Journal:J Biol Chem
PubMed ID:12000756
'Cysteine proteinases play a major role in invasion and intracellular survival of a number of pathogenic parasites. We cloned a single copy gene, tgcp1, from Toxoplasma gondii and refolded recombinant enzyme to yield active proteinase. Substrate specificity of the enzyme and homology modeling identified the proteinase as a cathepsin B. ... More
Misexpression of the eyes absent family triggers the apoptotic program.
Authors: Clark S Wesley; Fee Brian E; Cleveland John L;
Journal:J Biol Chem
PubMed ID:11700312
'Genetic studies in Drosophila and mice have shown that eyes absent (eya) is an important and conserved transcriptional regulator of development. Along with eyeless/Pax6, sine oculis, and dachshund, eya genes function as master regulators in eye development and can induce ectopic eye formation. Furthermore, the loss-of-function mutants of these genes ... More
Characterization of three alternatively spliced isoforms of the Rel/NF-kappa B transcription factor Relish from the mosquito Aedes aegypti.
Authors: Shin Sang Woon; Kokoza Vladimir; Ahmed Abduelaziz; Raikhel Alexander S;
Journal:Proc Natl Acad Sci U S A
PubMed ID:12119421
'The Rel/NF-kappa B transcription factor Relish performs a central role in the acute-phase response to microbial challenge by activating immune antibacterial peptides. We cloned and molecularly characterized the gene homologous to Drosophila Relish from the mosquito Aedes aegypti. Unlike Drosophila Relish, Aedes Relish has three alternatively spliced transcripts encoding different ... More