3-Phenoxybenzyl alcohol, 98%
3-Phenoxybenzyl alcohol, 98%
3-Phenoxybenzyl alcohol, 98%
Thermo Scientific Chemicals

3-Phenoxybenzyl alcohol, 98%

CAS: 13826-35-2 | C13H12O2 | 200.24 g/mol
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 183771000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13826-35-2
IUPAC Name(3-phenoxyphenyl)methanol
Molecular FormulaC13H12O2
InChI KeyKGANAERDZBAECK-UHFFFAOYSA-N
SMILESOCC1=CC=CC(OC2=CC=CC=C2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5910 to 1.5950 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only