How long does the labeling take when using the RadPrime DNA Labeling system?
Typically labeling is complete in 10 minutes, and purifiation of the probe is not usually required.
Search
View additional product information for RadPrime DNA Labeling System - FAQs (18428011)
1 product FAQs found